This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ACOX3 SIRNA Assays ›
Gene Symbol:
Acox3
Gene Name:
acyl-CoA oxidase 3, pristanoyl
Gene Aliases:
-
Chromosome Location:
Chr.14: 80313202 - 80358130 on Build Rnor_6.0
UniGene ID:
Rn.10546
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS331048
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Acox3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053339.1 NP_445791.1 17, 18 2017
XM_008770372.1 XP_008768594.1 17, 18 4777
XM_008770373.1 XP_008768595.1 17, 18 4820
XM_008770374.1 XP_008768596.1 16, 17 2044
GenBank mRNA X95188.1 - 17, 18 2017

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

fatty acid beta-oxidation
fatty acid beta-oxidation using acyl-CoA oxidase
acyl-CoA dehydrogenase activity
acyl-CoA oxidase activity
receptor binding
fatty acid binding
pristanoyl-CoA oxidase activity
flavin adenine dinucleotide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm