This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SERPIND1 SIRNA Assays ›
Gene Symbol:
Serpind1
Gene Name:
serpin peptidase inhibitor, clade D (heparin cofactor), member 1
Gene Aliases:
rls2var1
Chromosome Location:
Chr.11: 87913814 - 87924880 on Build Rnor_6.0
UniGene ID:
Rn.10553
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS330504
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Serpind1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024382.1 NP_077358.1 3 444
XM_006248709.2 XP_006248771.1 2 451
GenBank mRNA BC091145.1 - 3 424 7372906
FQ218607.1 - 2 421
X74549.1 - 3 444
X74550.1 - 2 391

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

blood coagulation
biological_process
negative regulation of endopeptidase activity
serine-type endopeptidase inhibitor activity
heparin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0