This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ANXA6 SIRNA Assays ›
Gene Symbol:
Anxa6
Gene Name:
annexin A6
Gene Aliases:
Anx6, CPB-II
Chromosome Location:
Chr.10: 40320581 - 40375605 on Build Rnor_6.0
UniGene ID:
Rn.204053
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS330471
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Anxa6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024156.2 NP_077070.2 12 958
XM_006246340.2 XP_006246402.1 12 1696
XM_006246341.2 XP_006246403.1 12 1697
GenBank mRNA BC072523.1 - 12 907 7105847
FQ219262.1 - 2 131

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

calcium ion transport
regulation of muscle contraction
ion transmembrane transport
secretion
protein homooligomerization
calcium ion binding
GTP binding
calcium-dependent phospholipid binding
lipid binding
ligand-gated ion channel activity
cholesterol binding
protein homodimerization activity
calcium-dependent protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0