This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ADRM1 SIRNA Assays ›
Gene Symbol:
Adrm1
Gene Name:
adhesion regulating molecule 1
Gene Aliases:
Gp110
Chromosome Location:
Chr.3: 175548060 - 175552943 on Build Rnor_6.0
UniGene ID:
Rn.9320
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS330231
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Adrm1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031708.1 NP_113896.1 5 559
XM_006235822.1 XP_006235884.1 5 680
GenBank mRNA AB032742.1 - 5 559
BC061773.1 - 5 575 5621873
FQ218007.1 - 5 577

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019822, BC008974, AK150091, AK145474, AF225959, and others.

Gene Ontology Categories:

Function(s) Process(es)

ovarian follicle development
transcription elongation from RNA polymerase II promoter
spermatid development
positive regulation of endopeptidase activity
regulation of T cell differentiation in thymus
follicle-stimulating hormone signaling pathway
proteasome assembly
oogenesis
thymus development
Sertoli cell development
positive regulation of growth hormone receptor signaling pathway
adipose tissue development
seminiferous tubule development
protease binding
endopeptidase activator activity
proteasome binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0