This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AQP3 SIRNA Assays ›
Gene Symbol:
Aqp3
Gene Name:
aquaporin 3
Gene Aliases:
-
Chromosome Location:
Chr.5: 57423735 - 57429252 on Build Rnor_6.0
UniGene ID:
Rn.11109
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS330226
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Aqp3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031703.1 NP_113891.1 3 326
GenBank mRNA BC127490.1 - 3 308 8366493
D17695.1 - 3 241
L35108.1 - 3 326

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of immune system process
water transport
cellular water homeostasis
glycerol transport
urea transport
response to retinoic acid
carbohydrate transmembrane transport
ion transmembrane transport
odontogenesis
renal water absorption
urea transmembrane transport
urea transmembrane transporter activity
water channel activity
glycerol channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0