This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AOC1 SIRNA Assays ›
Gene Symbol:
Aoc1
Gene Name:
amine oxidase, copper containing 1
Gene Aliases:
Abp, Abp1
Chromosome Location:
Chr.4: 78496043 - 78515585 on Build Rnor_6.0
UniGene ID:
Rn.54493
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS330159
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Aoc1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022935.2 NP_075224.2 2 258
XM_006236471.2 XP_006236533.1 2 280
XM_006236472.2 XP_006236534.1 2 101
XM_008762920.1 XP_008761142.1 2 282
GenBank mRNA BC100144.1 - 2 258 7460184
X73911.1 - 2 140

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cation transport
biological_process
amine metabolic process
cellular response to copper ion starvation
response to drug
response to antibiotic
oxidation-reduction process
cellular response to copper ion
cellular response to histamine
cellular response to azide
cation transmembrane transport
receptor activity
cation channel activity
copper ion binding
calcium ion binding
primary amine oxidase activity
drug binding
heparin binding
zinc ion binding
protein complex binding
protein homodimerization activity
quinone binding
diamine oxidase activity
histamine oxidase activity
methylputrescine oxidase activity
propane-1,3-diamine oxidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0