This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CHRNA9 SIRNA Assays ›
Gene Symbol:
Chrna9
Gene Name:
cholinergic receptor, nicotinic, alpha 9 (neuronal)
Gene Aliases:
-
Chromosome Location:
Chr.14: 43883450 - 43890171 on Build Rnor_6.0
UniGene ID:
Rn.54463
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS330150
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Chrna9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022930.1 NP_075219.1 2 188
GenBank mRNA AY574257.1 - 2 101
U12336.1 - 2 188

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cation transport
positive regulation of cytosolic calcium ion concentration
sensory perception of sound
inner ear morphogenesis
detection of mechanical stimulus involved in sensory perception of sound
calcium ion transmembrane transport
cation transmembrane transport
acetylcholine-activated cation-selective channel activity
calcium channel activity
acetylcholine-gated cation channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0