This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RBM10 SIRNA Assays ›
Gene Symbol:
Rbm10
Gene Name:
RNA binding motif protein 10
Gene Aliases:
-
Chromosome Location:
Chr.X: 1754869 - 1786973 on Build Rnor_6.0
UniGene ID:
Rn.8822
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS329958
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rbm10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152861.4 NP_690600.1 22 2763
GenBank mRNA D83948.1 - 22 2763

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
negative regulation of cell proliferation
regulation of smooth muscle cell apoptotic process
positive regulation of smooth muscle cell apoptotic process
negative regulation of mRNA splicing, via spliceosome
3'-UTR-mediated mRNA stabilization
nucleotide binding
zinc ion binding
protein complex binding
identical protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0