This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FHL2 SIRNA Assays ›
Gene Symbol:
Fhl2
Gene Name:
four and a half LIM domains 2
Gene Aliases:
-
Chromosome Location:
Chr.9: 49920611 - 49964902 on Build Rnor_6.0
UniGene ID:
Rn.3849
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS329644
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fhl2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031677.1 NP_113865.1 5 741
XM_006244849.2 XP_006244911.1 6 2878
GenBank mRNA AB008571.1 - 5 741

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010212, AK147024, AF153340, AF114381, AF055889, and others.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
osteoblast differentiation
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
response to hormone
negative regulation of apoptotic process
negative regulation of transcription, DNA-templated
atrial cardiac muscle cell development
ventricular cardiac muscle cell development
heart trabecula formation
protein binding
transcription factor binding
zinc ion binding
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header