This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ACACA SIRNA Assays ›
Gene Symbol:
Acaca
Gene Name:
acetyl-CoA carboxylase alpha
Gene Aliases:
ACC1, Acac
Chromosome Location:
Chr.10: 71519392 - 71719910 on Build Rnor_6.0
UniGene ID:
Rn.163753
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS329606
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Acaca
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022193.1 NP_071529.1 47 6365
GenBank mRNA EF121986.1 - 47 6365
EF121987.1 - 47 6365
J03808.1 - 47 6365

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_198839, NM_198834, NM_198836, NM_198837, NM_198838, and others.
This siRNA also targets the following transcripts in Mouse: NM_133360, AY451393, BC056500.

Gene Ontology Categories:

Function(s) Process(es)

tissue homeostasis
acetyl-CoA metabolic process
lipid metabolic process
fatty acid biosynthetic process
response to organic cyclic compound
response to drug
multicellular organismal protein metabolic process
protein homotetramerization
lipid homeostasis
cellular response to prostaglandin E stimulus
malonyl-CoA biosynthetic process
acetyl-CoA carboxylase activity
biotin carboxylase activity
ATP binding
biotin binding
kinase binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0