This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TCAM1 SIRNA Assays ›
Gene Symbol:
Tcam1
Gene Name:
testicular cell adhesion molecule 1
Gene Aliases:
Tcam1p
Chromosome Location:
Chr.10: 94465920 - 94478591 on Build Rnor_6.0
UniGene ID:
Rn.125075
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS329399
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tcam1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021673.1 NP_067705.1 4 748
XM_006247642.2 XP_006247704.1 5 1502
GenBank mRNA BC166495.1 - 5 810 7111123

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
single organismal cell-cell adhesion
integrin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header