This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CDKN2C SIRNA Assays ›
Gene Symbol:
Cdkn2c
Gene Name:
cyclin-dependent kinase inhibitor 2C (p18, inhibits CDK4)
Gene Aliases:
p18
Chromosome Location:
Chr.5: 129347731 - 129352886 on Build Rnor_6.0
UniGene ID:
Rn.63865
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS328966
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdkn2c
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_131902.3 NP_571977.1 809
GenBank mRNA AF474980.2 - 330
AF474981.2 - 311
BC088864.1 - 809 7319164

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cyclin-dependent protein serine/threonine kinase activity
G1/S transition of mitotic cell cycle
cell cycle arrest
negative regulation of cell proliferation
phosphorylation
negative regulation of cell growth
negative regulation of phosphorylation
oligodendrocyte differentiation
negative regulation of protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase inhibitor activity
kinase activity
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0