This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EDNRB SIRNA Assays ›
Gene Symbol:
Ednrb
Gene Name:
endothelin receptor type B
Gene Aliases:
Ednra, Etb
Chromosome Location:
Chr.15: 88004775 - 88036354 on Build Rnor_6.0
UniGene ID:
Rn.11412
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS328905
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ednrb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017333.1 NP_059029.1 6 1286
XM_006252431.2 XP_006252493.1 6 1567
GenBank mRNA S65355.1 - 6 1286
X57764.1 - 5 1235

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
neural crest cell migration
positive regulation of protein phosphorylation
regulation of pH
G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
peripheral nervous system development
posterior midgut development
aging
regulation of blood pressure
positive regulation of cell proliferation
negative regulation of neuron maturation
response to organic cyclic compound
vein smooth muscle contraction
sensory perception of pain
cGMP-mediated signaling
melanocyte differentiation
regulation of fever generation
negative regulation of cellular protein metabolic process
response to lipopolysaccharide
enteric smooth muscle cell differentiation
positive regulation of urine volume
positive regulation of renal sodium excretion
epithelial fluid transport
vasoconstriction
vasodilation
negative regulation of apoptotic process
pigmentation
developmental pigmentation
macrophage chemotaxis
response to pain
enteric nervous system development
regulation of epithelial cell proliferation
regulation of sensory perception of pain
positive regulation of penile erection
cellular response to lipopolysaccharide
endothelin receptor signaling pathway
receptor activity
G-protein coupled receptor activity
endothelin receptor activity
peptide hormone binding
type 1 angiotensin receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header