This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLR496 SIRNA Assays ›
Gene Symbol:
Olr496 LOC100912505
Gene Name:
olfactory receptor 496
olfactory receptor 8K3-like
Gene Aliases:
-
-
Chromosome Location:
Chr.3: 74789881 - 74790822 on Build Rnor_6.0
Chromosome Location:
Chr.3: 73676462 - 73677403 on Build Rnor_6.0
UniGene ID:
Rn.142946
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS328309
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr496
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000681.1 NP_001000681.1 1 293
Gene Symbol: LOC100912505
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749500.1 XP_003749548.1 1 293

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0