siRNA Details
| Gene Symbol: Olr537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000671.1 | NP_001000671.1 | 1 | 13 | ||

Olr537| Gene Symbol: Olr537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000671.1 | NP_001000671.1 | 1 | 13 | ||