This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RGD1310641 SIRNA Assays ›
Gene Symbol:
RGD1310641
Gene Name:
similar to hypothetical protein
Gene Aliases:
-
Chromosome Location:
Chr.5: 50148685 - 50193586 on Build Rnor_6.0
UniGene ID:
Rn.162804
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS327567
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RGD1310641
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079701.1 NP_001073169.1 7, 8 962
XM_006237999.2 XP_006238061.1 7, 8 1019
XM_008763628.1 XP_008761850.1 7, 8 875
XM_008763629.1 XP_008761851.1 6, 7 860
GenBank mRNA BC128759.1 - 7, 8 962 7112639

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0