This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TBL1XR1 SIRNA Assays ›
Gene Symbol:
Tbl1xr1
Gene Name:
transducin (beta)-like 1 X-linked receptor 1
Gene Aliases:
RGD1560053
Chromosome Location:
Chr.2: 107221987 - 107359229 on Build Rnor_6.0
UniGene ID:
Rn.140831
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS327265
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tbl1xr1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108941.1 NP_001102411.1 5 639
XM_006232190.2 XP_006232252.1 4 532
XM_008760889.1 XP_008759111.1 5 1395

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
response to dietary excess
regulation of transcription from RNA polymerase II promoter
biological_process
regulation of gene expression
lipid catabolic process
histone deacetylation
regulation of cAMP metabolic process
multicellular organism growth
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
white fat cell differentiation
canonical Wnt signaling pathway
adipose tissue development
fat pad development
regulation of triglyceride metabolic process
molecular_function
DNA binding
transcription corepressor activity
beta-catenin binding
histone binding
transcription regulatory region DNA binding
protein N-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0