This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRIM13 SIRNA Assays ›
Gene Symbol:
Trim13
Gene Name:
tripartite motif-containing 13
Gene Aliases:
-
Chromosome Location:
Chr.15: 41927180 - 41936529 on Build Rnor_6.0
UniGene ID:
Rn.83735
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS326613
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trim13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012210.1 NP_001012210.1 3 925
XM_006252137.2 XP_006252199.1 3 1012
XM_008770767.1 XP_008768989.1 3 993
XM_008770768.1 XP_008768990.1 2 909
GenBank mRNA BC088425.1 - 3 925 7123847

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
response to gamma radiation
positive regulation of cell death
positive regulation of macroautophagy
ER-associated ubiquitin-dependent protein catabolic process
negative regulation of viral transcription
positive regulation of I-kappaB kinase/NF-kappaB signaling
proteasome-mediated ubiquitin-dependent protein catabolic process
innate immune response
positive regulation of NF-kappaB transcription factor activity
protein autoubiquitination
negative regulation of viral release from host cell
ubiquitin-protein transferase activity
signal transducer activity
zinc ion binding
ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0