This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CEP85 SIRNA Assays ›
Gene Symbol:
Cep85
Gene Name:
centrosomal protein 85kDa
Gene Aliases:
Ccdc21, RGD1311520
Chromosome Location:
Chr.5: 152359693 - 152406528 on Build Rnor_6.0
UniGene ID:
Rn.20146
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS325264
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cep85
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080151.1 NP_001073620.1 7, 8 1414
XM_008764155.1 XP_008762377.1 7, 8 1495
XM_008764156.1 XP_008762378.1 7, 8 1489
XM_008764157.1 XP_008762379.1 5, 6 1626
GenBank mRNA BC129108.1 - 7, 8 1414 7104019

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0