This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP462 SIRNA Assays ›
Gene Symbol:
Zfp462
Gene Name:
zinc finger protein 462
Gene Aliases:
RGD1562123
Chromosome Location:
Chr.5: 71683931 - 71824077 on Build Rnor_6.0
UniGene ID:
-
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS325149
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp462
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006238178.2 XP_006238240.1 3 4610
XM_008763726.1 XP_008761948.1 2 9327
XM_008763727.1 XP_008761949.1 3 7315
XM_008763728.1 XP_008761950.1 2 9327
XM_008763729.1 XP_008761951.1 2 9327

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chromatin organization
negative regulation of DNA binding
positive regulation of transcription from RNA polymerase II promoter
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0