This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VPS26A SIRNA Assays ›
Gene Symbol:
Vps26a
Gene Name:
vacuolar protein sorting 26 homolog A (S. pombe)
Gene Aliases:
Vps26
Chromosome Location:
Chr.20: 32085465 - 32115310 on Build Rnor_6.0
UniGene ID:
Rn.3673
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS324310
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vps26a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007740.1 NP_001007741.1 7 761
XM_006256460.2 XP_006256522.1 7 884
XM_008772892.1 XP_008771114.1 7 889
GenBank mRNA BC079150.1 - 7 761 7128580
FQ211877.1 - 7 774

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001113355, NM_133672, AK028096, AK168213, AK165853, and others.

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
retrograde transport, endosome to Golgi
retrograde transport, endosome to plasma membrane
protein transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0