siRNA Details
Gene Symbol: Tmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108521.1 | NP_001101991.1 | 11 | 1405 |

More Information
Other Species Targeted

Gene Ontology Categories:

Gene Symbol: Tmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108521.1 | NP_001101991.1 | 11 | 1405 |