This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SYVN1 SIRNA Assays ›
Gene Symbol:
Syvn1
Gene Name:
synovial apoptosis inhibitor 1, synoviolin
Gene Aliases:
RGD1310488
Chromosome Location:
Chr.1: 221409265 - 221416225 on Build Rnor_6.0
UniGene ID:
Rn.162486
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS324147
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Syvn1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100739.1 NP_001094209.1 6 565
XM_006230852.2 XP_006230914.1 6 1007
GenBank mRNA BC166727.1 - 6 565 7933989

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
protein N-linked glycosylation via asparagine
ER-associated ubiquitin-dependent protein catabolic process
negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
zinc ion binding
acid-amino acid ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header