This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NAP1L4 SIRNA Assays ›
Gene Symbol:
Nap1l4
Gene Name:
nucleosome assembly protein 1-like 4
Gene Aliases:
-
Chromosome Location:
Chr.1: 216715352 - 216751738 on Build Rnor_6.0
UniGene ID:
Rn.3286
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS324112
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nap1l4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012170.1 NP_001012170.1 13, 14 1170
XM_006230823.2 XP_006230885.1 13, 14 1176
XM_006230824.2 XP_006230886.1 14, 15 1243
XM_006230826.2 XP_006230888.1 13, 14 1176
GenBank mRNA BC085801.1 - 13, 14 1170 7110617

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008672, NM_001285489, NM_001285490, BC145308, BC132039, and others.

Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0