This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RAD23A SIRNA Assays ›
Gene Symbol:
Rad23a
Gene Name:
RAD23 homolog A (S. cerevisiae)
Gene Aliases:
-
Chromosome Location:
Chr.19: 25949488 - 25956687 on Build Rnor_6.0
UniGene ID:
Rn.105419
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS323717
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rad23a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013190.1 NP_001013208.1 5 549
XM_006255276.2 XP_006255338.1 5 610
XM_006255277.2 XP_006255339.1 5 610
XM_008772411.1 XP_008770633.1 5 1865
GenBank mRNA BC084695.1 - 5 549 7114647

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001270362, NM_001270363, NM_005053, BC088364, AK289908, and others.

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair
cellular response to DNA damage stimulus
regulation of proteasomal ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of viral genome replication
damaged DNA binding
polyubiquitin binding
ubiquitin-specific protease binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0