This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TM9SF1 SIRNA Assays ›
Gene Symbol:
Tm9sf1
Gene Name:
transmembrane 9 superfamily member 1
Gene Aliases:
-
Chromosome Location:
Chr.15: 34314645 - 34322390 on Build Rnor_6.0
UniGene ID:
Rn.12071
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS323291
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tm9sf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012155.1 NP_001012155.1 1 646
XM_006251990.2 XP_006252052.1 2 486
XM_006251991.2 XP_006252053.1 2 502
XM_008770688.1 XP_008768910.1 1 703
GenBank mRNA BC081891.1 - 1 646 7109579

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028780, AK004754, AK090065, AK152540, BC007187, and others.

Gene Ontology Categories:

Function(s)

autophagy

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0