This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UCHL5 SIRNA Assays ›
Gene Symbol:
Uchl5
Gene Name:
ubiquitin carboxyl-terminal hydrolase L5
Gene Aliases:
-
Chromosome Location:
Chr.13: 60568109 - 60603812 on Build Rnor_6.0
UniGene ID:
Rn.40424
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS323062
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Uchl5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012149.1 NP_001012149.1 7 661
XM_006249960.2 XP_006250022.1 7 696
XM_006249961.2 XP_006250023.1 7 696
XM_006249962.2 XP_006250024.1 7 697
XM_006249963.2 XP_006250025.1 7 698
GenBank mRNA BC088841.1 - 7 661 7125859
FQ220301.1 - 7 667

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
biological_process
negative regulation of endopeptidase activity
protein deubiquitination
lateral ventricle development
midbrain development
forebrain morphogenesis
regulation of proteasomal protein catabolic process
molecular_function
thiol-dependent ubiquitin-specific protease activity
endopeptidase inhibitor activity
omega peptidase activity
poly(A) RNA binding
proteasome binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0