siRNA Details
Gene Symbol: Rhbdd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191827.1 | NP_001178756.1 | 4 | 941 | ||
XM_006249182.2 | XP_006249244.1 | 4 | 933 |

Gene Symbol: Rhbdd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191827.1 | NP_001178756.1 | 4 | 941 | ||
XM_006249182.2 | XP_006249244.1 | 4 | 933 |