This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RGD1310935 SIRNA Assays ›
Gene Symbol:
RGD1310935
Gene Name:
similar to Dermal papilla derived protein 7
Gene Aliases:
-
Chromosome Location:
Chr.11: 45713345 - 45763174 on Build Rnor_6.0
UniGene ID:
Rn.162005
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS322872
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RGD1310935
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008768658.1 XP_008766880.1 5, 6 1437
XM_008768659.1 XP_008766881.1 7, 8 996
XM_008768660.1 XP_008766882.1 6, 7 1353
XM_008768661.1 XP_008766883.1 5, 6 1407
GenBank mRNA FQ229246.1 - 3, 4 468

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0