This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC102550455 SIRNA Assays ›
Gene Symbol:
LOC102550455 Narf
Gene Name:
nuclear prelamin A recognition factor-like
nuclear prelamin A recognition factor
Gene Aliases:
-
RGD1310894
Chromosome Location:
Chr.10: 110125114 - 110128331 on Build Rnor_6.0
Chromosome Location:
Chr.10: 110445764 - 110463735 on Build Rnor_6.0
UniGene ID:
-
Rn.162225
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS322853
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC102550455
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006247957.2 XP_006248019.1 1, 2 537
Gene Symbol: Narf
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039207.2 NP_001034296.1 8, 9 930
XM_006247899.2 XP_006247961.1 8, 9 963
GenBank mRNA BC110050.1 - 8, 9 930 7454395

Back To Top

More Information


Gene Ontology Categories:

Process(es)

lamin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0