This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MYRIP SIRNA Assays ›
Gene Symbol:
Myrip
Gene Name:
myosin VIIA and Rab interacting protein
Gene Aliases:
Slac2c
Chromosome Location:
Chr.8: 128972383 - 129139257 on Build Rnor_6.0
UniGene ID:
Rn.161974
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS322578
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Myrip
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182844.1 NP_878264.1 13, 14 2356
XM_006244110.2 XP_006244172.1 13, 14 2396
XM_008766684.1 XP_008764906.1 14, 15 2474
XM_008766685.1 XP_008764907.1 15, 16 2588
XM_008766686.1 XP_008764908.1 13, 14 2398
XM_008766687.1 XP_008764909.1 12, 13 2869
GenBank mRNA AY331984.1 - 13, 14 2356

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
positive regulation of insulin secretion
actin binding
protein binding
myosin binding
Rab GTPase binding
metal ion binding
protein kinase A binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0