This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SHROOM2 SIRNA Assays ›
Gene Symbol:
Shroom2
Gene Name:
shroom family member 2
Gene Aliases:
Ab2-404, Apxl
Chromosome Location:
Chr.X: 23478869 - 23649427 on Build Rnor_6.0
UniGene ID:
Rn.16033
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS322403
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Shroom2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047893.3 NP_001041358.2 7 3264
XM_006256816.2 XP_006256878.1 7 3346
XM_006256817.2 XP_006256879.1 7 3343
XM_008773126.1 XP_008771348.1 6 3229
XM_008773127.1 XP_008771349.1 3 821
XM_008773128.1 XP_008771350.1 3 2489
XM_008773129.1 XP_008771351.1 3 2471
XM_008773130.1 XP_008771352.1 3 2474
XM_008773131.1 XP_008771353.1 3 803
XM_008773132.1 XP_008771354.1 3 806
GenBank mRNA AY325213.1 - 5 2955
CB586797.1 - 3 541
FQ211812.1 - 3 370

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172441, DQ358971, AK049850, CK635066, EF071946, and others.

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
multicellular organism development
cell migration
negative regulation of actin filament depolymerization
establishment of melanosome localization
melanosome organization
apical protein localization
cell-cell junction maintenance
actin filament bundle assembly
actin binding
beta-catenin binding
protein domain specific binding
actin filament binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header