This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DZIP1L SIRNA Assays ›
Gene Symbol:
Dzip1l
Gene Name:
DAZ interacting zinc finger protein 1-like
Gene Aliases:
RGD1311430
Chromosome Location:
Chr.8: 107875666 - 107916298 on Build Rnor_6.0
UniGene ID:
Rn.104060
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS321742
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dzip1l
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014095.1 NP_001014117.1 6 992
XM_006243626.2 XP_006243688.1 7 1196
XM_006243627.2 XP_006243689.1 7 1347
XM_008766526.1 XP_008764748.1 6 1960
XM_008766527.1 XP_008764749.1 6 1960
GenBank mRNA BC083786.1 - 6 992 7097259

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cilium assembly
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0