This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC4A8 SIRNA Assays ›
Gene Symbol:
Slc4a8
Gene Name:
solute carrier family 4, sodium bicarbonate cotransporter, member 8
Gene Aliases:
-
Chromosome Location:
Chr.7: 142397413 - 142464158 on Build Rnor_6.0
UniGene ID:
Rn.210164
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS321321
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc4a8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270794.1 NP_001257723.1 11 1392
NM_001270795.1 NP_001257724.1 11 1392
NM_199497.2 NP_955791.1 11 1392
GenBank mRNA AY489024.1 - 11 1392
AY489025.1 - 11 1392
AY489026.1 - 11 1392

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

bicarbonate transport
sodium ion transmembrane transport
regulation of intracellular pH
anion transmembrane transport
inorganic anion exchanger activity
sodium:bicarbonate symporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0