This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MKL1 SIRNA Assays ›
Gene Symbol:
Mkl1
Gene Name:
megakaryoblastic leukemia (translocation) 1
Gene Aliases:
BSAC, FLMKL1, MELODY, MKL1met, Mrtf-a
Chromosome Location:
Chr.7: 122233196 - 122403780 on Build Rnor_6.0
UniGene ID:
Rn.106726
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS321232
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mkl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750385.3 XP_003750433.1 3, 4 5896
XM_006242130.2 XP_006242192.1 4, 5 953
XM_008765806.1 XP_008764028.1 3, 4 1566
XM_235497.8 XP_235497.6 4, 5 627
GenBank mRNA AB588919.1 - 4, 5 514
AB588920.1 - 3, 4 369
AB588921.1 - 4, 5 496

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_153049, NM_001082536, BB850443, AF385582, AK155821, and others.

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
positive regulation of transcription via serum response element binding
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
smooth muscle cell differentiation
negative regulation of apoptotic signaling pathway
transcription regulatory region sequence-specific DNA binding
molecular_function
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
actin binding
actin monomer binding
leucine zipper domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0