This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRPF39 SIRNA Assays ›
Gene Symbol:
Prpf39
Gene Name:
pre-mRNA processing factor 39
Gene Aliases:
-
Chromosome Location:
Chr.6: 86785793 - 86810806 on Build Rnor_6.0
UniGene ID:
Rn.12521
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS320655
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prpf39
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750171.2 XP_003750219.1 6 965
XM_006240140.2 XP_006240202.1 6 1473
XM_006240141.2 XP_006240203.1 7 1370
XM_006240142.2 XP_006240204.1 6 1035
XM_006240143.2 XP_006240205.1 6 683
XM_006240144.2 XP_006240206.1 5 619
GenBank mRNA BC166472.1 - 5 1595

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_177806, BC030904, AK168462, BC014689, AK154170.

Gene Ontology Categories:

Function(s)

mRNA processing

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0