This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SPSB1 SIRNA Assays ›
Gene Symbol:
Spsb1
Gene Name:
splA/ryanodine receptor domain and SOCS box containing 1
Gene Aliases:
RGD1309319
Chromosome Location:
Chr.5: 166913734 - 166978495 on Build Rnor_6.0
UniGene ID:
Rn.219023
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS320433
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Spsb1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107994.2 NP_001101464.1 2 786
XM_006239462.2 XP_006239524.1 2 819
XM_008764257.1 XP_008762479.1 2 1032
XM_008764258.1 XP_008762480.1 1 617
XM_008764259.1 XP_008762481.1 2 2763
GenBank mRNA BC158603.1 - 2 786 7133644

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029035, AK030417, AK003707, BC089357, AK089767, and others.

Gene Ontology Categories:

Function(s)

intracellular signal transduction

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0