This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZMYM1 SIRNA Assays ›
Gene Symbol:
Zmym1
Gene Name:
zinc finger, MYM-type 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 145106518 - 145123869 on Build Rnor_6.0
UniGene ID:
Rn.19963
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS320356
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zmym1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107983.1 NP_001101453.1 8 3180
XM_006238878.2 XP_006238940.1 6 4384
XM_006238879.1 XP_006238941.1 8 3094
XM_006238880.2 XP_006238942.1 7 3118
GenBank mRNA BC160845.1 - 7 3073
FQ219503.1 - 1 829

Back To Top

More Information


Gene Ontology Categories:

Process(es)

nucleic acid binding
zinc ion binding
protein dimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0