This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CECR5 SIRNA Assays ›
Gene Symbol:
Cecr5
Gene Name:
cat eye syndrome chromosome region, candidate 5
Gene Aliases:
-
Chromosome Location:
Chr.4: 153047233 - 153079069 on Build Rnor_6.0
UniGene ID:
Rn.18084
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS319826
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cecr5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107884.1 NP_001101354.1 2 154
XM_006237246.2 XP_006237308.1 2 617
XM_006237247.2 XP_006237309.1 2 619

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_144815, BC014705, AK140387, AK044228.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0