This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PLXND1 SIRNA Assays ›
Gene Symbol:
Plxnd1
Gene Name:
plexin D1
Gene Aliases:
-
Chromosome Location:
Chr.4: 147854309 - 147894173 on Build Rnor_6.0
UniGene ID:
Rn.21111
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS319798
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Plxnd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107881.1 NP_001101351.1 3 1386
XM_006237035.2 XP_006237097.1 3 1654

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_015103, CX761094, AY116661, BC150280, AB014520.

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
patterning of blood vessels
outflow tract morphogenesis
synapse assembly
regulation of cell migration
positive regulation of protein binding
endothelial cell migration
regulation of angiogenesis
dichotomous subdivision of terminal units involved in salivary gland branching
semaphorin-plexin signaling pathway
semaphorin receptor activity
protein domain specific binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0