This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRC8A SIRNA Assays ›
Gene Symbol:
Lrrc8a
Gene Name:
leucine rich repeat containing 8 family, member A
Gene Aliases:
Lrrc8
Chromosome Location:
Chr.3: 8802852 - 8829503 on Build Rnor_6.0
UniGene ID:
Rn.33512
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS319491
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lrrc8a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024782.1 NP_001019953.1 3 335
XM_006233814.2 XP_006233876.1 1 181
XM_008761660.1 XP_008759882.1 1 181
GenBank mRNA BC097371.1 - 3 335 7444090

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_177725, BQ745966, BQ444045.

Gene Ontology Categories:

Function(s) Process(es)

pre-B cell differentiation
anion transport
cell volume homeostasis
response to osmotic stress
biological_process
anion transmembrane transport
molecular_function
anion channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0