siRNA Details
Gene Symbol: Gstcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107725.1 | NP_001101195.1 | 12 | 2020 | ||
XM_008761505.1 | XP_008759727.1 | 11 | 3503 | |||
GenBank mRNA | FQ211741.1 | - | 11 | 1974 |
Gene Symbol: LOC102556234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233344.2 | XP_006233406.1 | 608 |
