siRNA Details
Gene Symbol: Fam105a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037648.1 | NP_001032737.1 | 5 | 439 | ||
GenBank mRNA | BC107649.1 | - | 5 | 439 | 7930470 |

Gene Symbol: Fam105a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037648.1 | NP_001032737.1 | 5 | 439 | ||
GenBank mRNA | BC107649.1 | - | 5 | 439 | 7930470 |