This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PDE8B SIRNA Assays ›
Gene Symbol:
Pde8b
Gene Name:
phosphodiesterase 8B
Gene Aliases:
RNPDE8B
Chromosome Location:
Chr.2: 24718548 - 24923718 on Build Rnor_6.0
UniGene ID:
Rn.69724
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS318318
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pde8b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199268.1 NP_954889.1 4 357
XM_006231811.2 XP_006231873.1 5 971
XM_008760669.1 XP_008758891.1 5 968
XM_008760670.1 XP_008758892.1 5 971
GenBank mRNA AB092697.1 - 4 357

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

phosphorelay signal transduction system
behavioral fear response
metabolic process
visual learning
operant conditioning
negative regulation of insulin secretion
neuromuscular process controlling balance
negative regulation of steroid hormone biosynthetic process
molecular_function
3',5'-cyclic-AMP phosphodiesterase activity
signal transducer activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0