This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CHUK SIRNA Assays ›
Gene Symbol:
Chuk
Gene Name:
conserved helix-loop-helix ubiquitous kinase
Gene Aliases:
Ikbka, Ikka
Chromosome Location:
Chr.1: 263848829 - 263884354 on Build Rnor_6.0
UniGene ID:
Rn.23019
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS317959
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Chuk
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107588.1 NP_001101058.1 7 738
XM_006231481.2 XP_006231543.1 7 735
XM_008760418.1 XP_008758640.1 7 736
XM_008760419.1 XP_008758641.1 7 736
GenBank mRNA BC168905.1 - 7 713 9088878

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007700, NM_001162410, AK164236, AK035326, AK018671, and others.

Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of an epithelial sheet
skeletal muscle contraction
protein phosphorylation
I-kappaB phosphorylation
Rho protein signal transduction
lactation
response to toxic substance
response to organic substance
response to acetate
osteoclast differentiation
response to lipopolysaccharide
response to hydroperoxide
mammary gland epithelial cell proliferation
response to muscle stretch
odontogenesis of dentin-containing tooth
response to drug
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to amino acid
positive regulation of transcription from RNA polymerase II promoter
striated muscle cell differentiation
mammary gland alveolus development
cellular response to tumor necrosis factor
protein kinase activity
ATP binding
IkappaB kinase activity
protein homodimerization activity
protein heterodimerization activity
scaffold protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0