This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SMPD1 SIRNA Assays ›
Gene Symbol:
Smpd1
Gene Name:
sphingomyelin phosphodiesterase 1, acid lysosomal
Gene Aliases:
-
Chromosome Location:
Chr.1: 170383682 - 170387525 on Build Rnor_6.0
UniGene ID:
Rn.18277
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS317615
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Smpd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006997.1 NP_001006998.1 6 1799
GenBank mRNA BC083780.1 - 6 1799 7096039

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001007593, NM_000543, X59960, X52679, AK292388, and others.
This siRNA also targets the following transcripts in Mouse: NM_011421, AK206661, AK145702, AK145534, AK164167, and others.

Gene Ontology Categories:

Function(s) Process(es)

sphingomyelin catabolic process
termination of signal transduction
positive regulation of protein dephosphorylation
response to cocaine
response to drug
positive regulation of apoptotic process
negative regulation of MAP kinase activity
ceramide biosynthetic process
molecular_function
sphingomyelin phosphodiesterase activity
hydrolase activity, acting on glycosyl bonds

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0