This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP383 SIRNA Assays ›
Gene Symbol:
Zfp383 LOC103690114
Gene Name:
zinc finger protein 383
zinc finger protein 383-like
Gene Aliases:
-
-
Chromosome Location:
Chr.1: 88204835 - 88216548 on Build Rnor_6.0
Chromosome Location:
Chr.1: 87790111 - 87801824 on Build Rnor_6.0
UniGene ID:
-
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS317314
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp383
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006228746.2 XP_006228808.1 831
XM_006228747.2 XP_006228809.1 763
XM_006228748.2 XP_006228810.1 878
XM_006228749.2 XP_006228811.1 716
XM_006228750.2 XP_006228812.1 934
XM_006228751.2 XP_006228813.1 653
Gene Symbol: LOC103690114
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006228705.2 XP_006228767.1 4 831
XM_006228706.2 XP_006228768.1 5 763
XM_006228707.2 XP_006228769.1 5 878
XM_006228708.2 XP_006228770.1 5 716
XM_006228709.2 XP_006228771.1 5 934
XM_006228710.2 XP_006228772.1 5 653

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
nucleic acid binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0