This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC7A6 SIRNA Assays ›
Gene Symbol:
Slc7a6
Gene Name:
solute carrier family 7 (amino acid transporter light chain, y+L system), member 6
Gene Aliases:
-
Chromosome Location:
Chr.19: 38152225 - 38179149 on Build Rnor_6.0
UniGene ID:
Rn.8066
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS316898
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc7a6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107424.1 NP_001100894.1 6 1142
XM_006255498.2 XP_006255560.1 7 1207
XM_006255499.2 XP_006255561.1 7 1237
XM_006255500.2 XP_006255562.1 7 1355
XM_008772500.1 XP_008770722.1 7 1241
XM_008772501.1 XP_008770723.1 7 1422
XM_008772502.1 XP_008770724.1 7 1471

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
amino acid transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0