This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DCAF11 SIRNA Assays ›
Gene Symbol:
Dcaf11
Gene Name:
DDB1 and CUL4 associated factor 11
Gene Aliases:
Wdr23
Chromosome Location:
Chr.15: 34233851 - 34244512 on Build Rnor_6.0
UniGene ID:
Rn.16542
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS315902
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dcaf11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009686.2 NP_001009686.1 11 1249
XM_006251971.2 XP_006252033.1 12 1584
GenBank mRNA BC081957.1 - 12 1454
BC087110.1 - 10 1030 7193336
BC091364.1 - 11 1249 7319156
FQ222851.1 - 12 1550

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133734, NM_001199009, AK152395, BC037001, AK033540, and others.

Gene Ontology Categories:

Function(s)

protein ubiquitination

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0