This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM53A SIRNA Assays ›
Gene Symbol:
Fam53a
Gene Name:
family with sequence similarity 53, member A
Gene Aliases:
RGD1563315
Chromosome Location:
Chr.14: 82375093 - 82416004 on Build Rnor_6.0
UniGene ID:
Rn.19744
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS315673
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam53a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107228.2 NP_001100698.1 3, 4 778
XM_006251262.2 XP_006251324.1 3, 4 839
XM_006251266.2 XP_006251328.1 2, 3 740
XM_006251267.2 XP_006251329.1 2, 3 788
XM_006251268.2 XP_006251330.1 2, 3 713
XM_006251270.2 XP_006251332.1 2, 3 253
XM_008770286.1 XP_008768508.1 2, 3 1226
XM_008770287.1 XP_008768509.1 2, 3 1230
XM_008770288.1 XP_008768510.1 2, 3 257
GenBank mRNA BC168895.1 - 3, 4 778 7312953

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0